Anti-HBA1 Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 7E2]

Leverandør: Abnova
MAB0030-M07
ABNOMAB0030-M07EA 4800 NOK
ABNOMAB0030-M07
Anti-HBA1 Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 7E2]
Antistoffer
Mouse monoclonal antibody raised against glycosylated hemoglobin.

Type: Primary
Antigen: HBA1
Clonality: Monoclonal
Clone: 7E2
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR