Anti-NCAM1/795 Mouse Monoclonal Antibody [clone: NCAM1/795]

Leverandør: Abnova
MAB13381
ABNOMAB13381EA 6125 NOK
ABNOMAB13381
Anti-NCAM1/795 Mouse Monoclonal Antibody [clone: NCAM1/795]
Antistoffer
Mouse monoclonal antibody raised against full length recombinant human NCAM1.

Type: Primary
Antigen: NCAM1/795
Clonality: Monoclonal
Clone: NCAM1/795
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR