Anti-Gentamicin Monoclonal Antibody [clone: 73.3]

Leverandør: Abnova
MAB2226
ABNOMAB2226EA 4725 NOK
ABNOMAB2226
Anti-Gentamicin Monoclonal Antibody [clone: 73.3]
Antistoffer
Mouse monoclonal antibody raised against Gentamicin.

Type: Primary
Antigen: Gentamicin
Clonality: Monoclonal
Clone: 73.3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR