Anti-Gentamicin Monoclonal Antibody [clone: 26.16]

Leverandør: Abnova
MAB2227
ABNOMAB2227EA 4725 NOK
ABNOMAB2227
Anti-Gentamicin Monoclonal Antibody [clone: 26.16]
Antistoffer
Mouse monoclonal antibody raised against Gentamicin.

Type: Primary
Antigen: Gentamicin
Clonality: Monoclonal
Clone: 26.16
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR