Anti-JAK3 (pTyr980/981) Rabbit Monoclonal Antibody [clone: E10]

Leverandør: Abnova
MAB23565.200UL
ABNOMAB23565.200ULEA 10675 NOK
ABNOMAB23565.200UL
Anti-JAK3 (pTyr980/981) Rabbit Monoclonal Antibody [clone: E10]
Antistoffer
Bestill nå

SPESIFIKASJONER

Antigensymbol JAK3
Environmentally Preferable
Klonalitet Monoclonal
Klon E10
Reaktivitet Human
Vert Rabbit
Isotype
Modifikasjon pTyr980/981
Immunogen Rabbit monoclonal antibody raised against synthetic phosphopeptide of human JAK3.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR