Anti-KLH Polyclonal Antibody

Leverandør: Abnova
PAB0898
ABNOPAB0898EA 4725 NOK
ABNOPAB0898
Anti-KLH Polyclonal Antibody
Antistoffer
Rabbit polyclonal antibody raised against Keyhole Limpet Hemocyanin, KLH.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: KLH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR