Anti-8-epi-PGF2-alpha Polyclonal Antibody

Leverandør: Abnova
PAB13606
ABNOPAB13606EA 6125 NOK
ABNOPAB13606
Anti-8-epi-PGF2-alpha Polyclonal Antibody
Antistoffer
Goat polyclonal antibody raised against 8-epi-PGF2-alpha.

Type: Primary
Antigen: 8-epi-PGF2-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR