Anti-SLC7A11 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB14939
ABNOPAB14939EA 9125 NOK
ABNOPAB14939
Anti-SLC7A11 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of Slc7a11.

Type:
Antigen: SLC7A11
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR