Anti-SYCP1 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB14959
ABNOPAB14959EA 9125 NOK
ABNOPAB14959
Anti-SYCP1 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of SYCP1.

Recommended Dilutions: Immunofluorescence: 1:300-1:750

Type:
Antigen: SYCP1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR