Anti-SPECC1 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB14982
ABNOPAB14982EA 9125 NOK
ABNOPAB14982
Anti-SPECC1 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of CYTSB.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type:
Antigen: SPECC1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR