Anti-LYVE1 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB14998
ABNOPAB14998EA 10675 NOK
ABNOPAB14998
Anti-LYVE1 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of Lyve1.

Recommended Dilutions: Flow Cytometry: 1:25

Type:
Antigen: LYVE1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR