Anti-IREB2 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15011
ABNOPAB15011EA 9125 NOK
ABNOPAB15011
Anti-IREB2 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of Ireb2.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type:
Antigen: IREB2
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR