Anti-CTGF Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15038
ABNOPAB15038EA 9125 NOK
ABNOPAB15038
Anti-CTGF Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of CTGF.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type:
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR