Anti-TUBB Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15099
ABNOPAB15099EA 9125 NOK
ABNOPAB15099
Anti-TUBB Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of TUBB.

Recommended Dilutions: Western Blot: 1:1000-1:10000

Type:
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Pig, Rat, Xenopus, Zebrafish
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR