Anti-TRPM2 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15106
ABNOPAB15106EA 9125 NOK
ABNOPAB15106
Anti-TRPM2 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of Trpm2.

Recommended Dilutions: Immunohistochemistry: 1:200; Western Blot: 1:500-1:1000

Type:
Antigen: TRPM2
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR