Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight 549)

Leverandør: Abnova
PAB15114
ABNOPAB15114EA 10675 NOK
ABNOPAB15114
Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of TRPA1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:250

Type:
Antigen: TRPA1
Clonality: DyLight 549
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human, Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol TRPA1
Antigen-navn transient receptor potential cation channel, subfamily A, member 1
Environmentally Preferable
Antigen synonymer ANKTM1|FEPS
Klonalitet Polyclonal
Konjugering DyLight® 549
Reaktivitet Human,Mouse
Vert Rabbit
Gen-ID 8989
Isotype
Western blot Yes
ImmunoKjemi Yes
Immunogen A synthetic peptide corresponding to amino acids 1-100 of human TRPA1.
Aminosyretall 1 to 100
Format Liquid
Kyssabsorpsjon No
Lagringsbuffer 50 mM sodium borate
Lagringstemp Store at 4 °C. Do not freeze.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR