Anti-TERF2 Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15119
ABNOPAB15119EA 10675 NOK
ABNOPAB15119
Anti-TERF2 Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against recombinant TERF2.

Recommended Dilutions: ChIP: 1:2000-1:5000; Immunofluorescence: 1:2000-1:5000; Western Blot: 1:2000-1:5000

Type:
Antigen: TERF2
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host:
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR