Anti-TMED10 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15137
ABNOPAB15137EA 9125 NOK
ABNOPAB15137
Anti-TMED10 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of TMED10.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry: 1 ?g/ml

Type:
Antigen: TMED10
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Primates, Rabbit, Rat, Xenopus
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR