Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15147
ABNOPAB15147EA 9125 NOK
ABNOPAB15147
Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of Plin3.

Recommended Dilutions: Western Blot: 1:1000-1:5000

Type:
Antigen: PLIN3
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR