Anti-SPDYA Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15207
ABNOPAB15207EA 9125 NOK
ABNOPAB15207
Anti-SPDYA Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of SPDYA.

Type:
Antigen: SPDYA
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol SPDYA
Environmentally Preferable
Antigen-navn Speedy Homolog A
Antigen synonymer RINGOA|SPDY1|RINGO3|SPY1
Klonalitet Polyclonal
Konjugering DyLight® 549
Reaktivitet Mouse
Vert Rabbit
Gen-ID 245711
Isotype
Western blot Yes
Immunogen A synthetic peptide corresponding to amino acids 200-286 of human SPDYA.
Aminosyretall 200 to 286
Format Liquid
Kyssabsorpsjon No
Lagringsbuffer 50 mM sodium borate
Lagringstemp Store at 4 °C. Do not freeze.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR