Anti-SYCP3 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15246
ABNOPAB15246EA 9125 NOK
ABNOPAB15246
Anti-SYCP3 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of Sycp3.

Recommended Dilutions: Immunofluorescence: 1:100-1:750; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:750

Type:
Antigen: SYCP3
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR