Anti-ROBO4 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15258
ABNOPAB15258EA 9125 NOK
ABNOPAB15258
Anti-ROBO4 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of ROBO4.

Recommended Dilutions: Western Blot: 1:500; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5 ?g/ml

Type:
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol ROBO4
Environmentally Preferable
Antigen-navn Roundabout Homolog 4
Antigen synonymer MRB|MGC133352|MGC133353|UNQ421/PRO3674|FLJ20798|ECSM4
Klonalitet Polyclonal
Konjugering DyLight® 549
Reaktivitet Human
Vert Rabbit
Gen-ID 54538
Isotype
Western blot Yes
ImmunoKjemi Yes
Immunogen A synthetic peptide corresponding to amino acids 900-1007 of human ROBO4.
Aminosyretall 900 to 1007
Format Liquid
Kyssabsorpsjon No
Lagringsbuffer 50 mM sodium borate
Lagringstemp Store at 4 °C. Do not freeze.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR