Anti-PDPK1 Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15332
ABNOPAB15332EA 9125 NOK
ABNOPAB15332
Anti-PDPK1 Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of PDK1.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type:
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR