Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 549)

Leverandør: Abnova
PAB15353
ABNOPAB15353EA 9125 NOK
ABNOPAB15353
Anti-CDKN2A Rabbit Polyclonal Antibody (DyLight® 549)
Antistoffer
Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A.

Recommended Dilutions: Immunofluorescence: 1:100; Western Blot: 2 ?g/ml

Type:
Antigen: CDKN2A
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR