Avantor® ACE® SuperC18, HPLC Columns, 5 µm

Leverandør: Avantor
ACE-1211-1010EA 16975 NOK
ACE-1211-1010 ACE-1211-1020 ACE-1211-1220 ACE-1211-1510 ACE-1211-1520 ACE-1211-2510 ACE-1211-2520 ACE-1211-0510 ACE-1211-0520 ACE-1211-2530 ACE-1211-15005 ACE-1211-0103GD ACE-1211-0102GD ACE12110110GD ACE12110120GD ACE-1211-0530 ACE-1211-7530 ACE-1211-1030 ACE-1211-1530 ACE-1211-3030
Avantor® ACE® SuperC18, HPLC Columns, 5 µm
Kromatografikolonner
Avantor® ACE® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.

  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1,7 to 10 µm) and complementary phases

Maximum recommended pH: 1,5 to 11,5

Max. pressure: 275 bar
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Karbonbelastning 14,8% 14,8%
Kappet ved endene Ja Yes
Maks. trykk 275 bar
pH område 1,5 - 11,5 1,5 - 11,5
Overflate areal 400 m²/g 400 m²/g
USP kode L1 L1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR