Avantor® ACE® NH₂, HPLC Columns, 5 µm
Leverandør: Avantor
ACE-1214-2508EA 18275 NOK
ACE-1214-2508 ACE-1214-2510 ACE-1214-2520 ACE-1214-2530 ACE-1214-3030 ACE-1214-0510 ACE-1214-0520 ACE-1214-0530 ACE-1214-7520 ACE-1214-7530 ACE-1214-1010 ACE-1214-1020 ACE-1214-1030 ACE-1214-1220 ACE-1214-1510 ACE-1214-1520 ACE-1214-1530
Avantor® ACE® NH₂, HPLC Columns, 5 µm
Kromatografikolonner
These stainless steel columns with proprietary bonding technology provide high robustness with excellent column lifetimes.
  • Usable in reversed phase, normal phase, weak anion-exchange and HILIC modes
  • Ideal for the analysis of sugars
  • Ultra-inert, high efficiency with optimum peak shape
  • Range of particle sizes: 1,7 µm, 3 µm and 5 µm and complementary phases