Avantor® ACE®, HPLC ¹/₃₂" Column Connectors, Finger-Tight
Leverandør: Avantor
ACE-MC3210EA 0 NOK
ACE-MC3210
Avantor® ACE®, HPLC ¹/₃₂" Column Connectors, Finger-Tight
Fittings og kontakter
PEEK ¹/₃₂" microbore HPLC column inlet / outlet connectors. These column connectors are recommended for ¹/₃₂" microbore HPLC column connections. They are compatible with both capillary (360 µm OD) and ¹/₃₂" connection tubing.
  • Pressure rated to 400 bar (6000 psi)
  • Reusable
  • Compatible with all microbore columns with ¹/₃₂" and 6-40 threads