N-Acetylglycine(Ac-Gly-OH) 99%

Leverandør: Thermo Fisher Scientific

102571000 102575000
ACRO102571000EA 375 NOK
ACRO102571000 ACRO102575000
N-Acetylglycine(Ac-Gly-OH) 99%
N-Acetylglycine(Ac-Gly-OH)
Formel: C₄H₇NO₃
Smeltepunkt: 207…209 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004275
CAS-nummer: 543-24-8
EINECS: 208-839-6

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Titration with NaOH ≥98.5 % (On dry substance)
Impurity ≤0.25 % Glycine
Sulfate (SO4) ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR