ɑ-Amino-γ-butyrolactone hydrobromide 99%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: DL-Homoserine lactone hydrobromide

103290100 103290025 103290500
ACRO103290100EA 2300 NOK
ACRO103290100 ACRO103290025 ACRO103290500
ɑ-Amino-γ-butyrolactone hydrobromide 99%
alpha-Amino-gamma-butyrolactone hydrobromide

CAS No.: 6305-38-0

Formel: C₄H₇NO₂
MW: 182.02 g/mol
Smeltepunkt: 221 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012723
CAS-nummer: 6305-38-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Dye content ≥65 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR