L(+)-Arabinose 99+%

Leverandør: Thermo Fisher Scientific

104980250 104981000 104985000
ACRO104980250EA 870 NOK
ACRO104980250 ACRO104981000 ACRO104985000
L(+)-Arabinose 99+%
L(+)-Arabinose
Formel: C₅H₁₀O₅
Kokepunkt: 331 °C (1013 hPa)
Smeltepunkt: 155…160 °C
Tetthet: 1,585 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006609
CAS-nummer: 87-72-9
EINECS: 226-214-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.3 %
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation after 24 h
Sodium (Na) ≤0.2 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR