Benzhydryl alcohol 99%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Diphenylcarbinol, Benzhydrol, Diphenylmethanol

105391000 105395000
ACRO105391000EA 510 NOK
ACRO105391000 ACRO105395000
Benzhydryl alcohol 99%
Benzhydryl alcohol
Formel: C₁₃H₁₂O
MW: 184.24 g/mol
Kokepunkt: 297
Smeltepunkt: 65…68 °C
Flash punkt: 160 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004488
CAS-nummer: 91-01-0
EINECS: 202-033-8
Merck Indeks: 12,01121

Bestill nå

Spesifikasjon Testresultater

Appearance White to beige Crystalline solid
Infrared spectrum Conforms
Melting point 65 to 68 °C
Water ≤0.5 % (K.F.)
Loss on drying ≤0.5 % (55 °C)
GC ≥98.5 %
Lead (Pb) ≤10 ppm
Nickel (Ni) ≤100 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR