Diethyl chlorophosphite 90% tech.

Leverandør: Thermo Fisher Scientific
Danger

114250250 114251000
ACRO114250250EA 1675 NOK
ACRO114250250 ACRO114251000
Diethyl chlorophosphite 90% tech.
Diethyl chlorophosphite
Formel: C₄H₁₀ClO₂P
Tetthet: 1,09 g/cm³
Flash punkt: <1 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009074
CAS-nummer: 589-57-1
EINECS: 209-652-2
FN: 2924
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4410 (20°C, 589 nm)
Titration Argentometric ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR