N,N-Dimetyl-p-toluidine 99%

Leverandør: Thermo Fisher Scientific
Danger

116905000 116900025
ACRO116905000EA 1925 NOK
ACRO116905000 ACRO116900025
N,N-Dimetyl-p-toluidine 99%
N,N-Dimetyl-p-toluidine
Formel: C₉H₁₃N
Kokepunkt: 211 °C (760 mmHg)
Smeltepunkt: -15 °C
Tetthet: 0,93 g/cm³
Flash punkt: 83 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008316
CAS-nummer: 99-97-8
EINECS: 202-805-4
FN: 2810
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Color scale ≤10 APHA
Stabilizer 0.5 to 1.0 % Ethanol
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Free chlorine (Cl) Passes test
Lead (Pb) ≤0.05 ppm
Acid and chloride Passes test
Acetone and aldehyde Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR