Jern 98%

Leverandør: Thermo Fisher Scientific
Warning

119141000 119145000 119140025
ACRO119141000EA 1000 NOK
ACRO119141000 ACRO119145000 ACRO119140025
Jern 98%
Jern
Formel: C₁₀H₁₀Fe
Kokepunkt: 249 °C (760 mmHg)
Smeltepunkt: 173…176 °C
Tetthet: 1,49 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001427
CAS-nummer: 102-54-5
EINECS: 203-039-3
FN: 1325
ADR: 4.1,II
Merck Indeks: 13,04070

Bestill nå

Spesifikasjon Testresultater

Appearance Orange-brown Crystalline powder or crystals (May become lumpy on storage)
Infrared spectrum Conforms
Melting point 173 to 176 °C
GC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR