Fisetin

Leverandør: Thermo Fisher Scientific
Warning

119150010 119150050
ACRO119150010EA 2200 NOK
ACRO119150010 ACRO119150050
Fisetin
Fisetin
Formel: C₁₅H₁₀O₆
Smeltepunkt: >330 °C
MDL nummer: MFCD00006829
CAS-nummer: 528-48-3
EINECS: 208-434-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to amber to dark green Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR