1-Heksadeken 92% tech.

Leverandør: Thermo Fisher Scientific
Danger

120500010
ACRO120500010EA 1575 NOK
ACRO120500010
1-Heksadeken 92% tech.
1-Heksadeken
Formel: C₁₆H₃₂
Kokepunkt: 285 °C (1013 hPa)
Smeltepunkt: 3…5 °C
Tetthet: 0,783 g/cm³
Flash punkt: 132 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008991
CAS-nummer: 629-73-2
EINECS: 211-105-8

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20°C, 589 nm)
GC ≥90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR