4-Quinazolone 98%

Leverandør: Thermo Fisher Scientific
Warning

121880250
ACRO121880250EA 1100 NOK
ACRO121880250
4-Quinazolone 98%
4-Quinazolone
Formel: C₈H₆N₂O
Smeltepunkt: 215…219 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00511302
CAS-nummer: 491-36-1
EINECS: 207-735-8

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 215 to 219 °C
Titration with TBAH ≥97.5 %
Enantiomeric excess ≥97.5 %
Specific optical rotation -118° to -122° (20 °C, 589 nm) (c=5, in CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR