Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene/methanol for titration in non-aqueous media

Leverandør: Thermo Fisher Scientific
Danger

123531000 123535000 123530025
ACRO123531000EA 815 NOK
ACRO123531000 ACRO123535000 ACRO123530025
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene/methanol for titration in non-aqueous media
Tetrabutylammonium hydroxide
Formel: C₁₆H₃₇NO
Tetthet: 0,85 g/cm³
Flash punkt: 4 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009425
CAS-nummer: 2052-49-5
EINECS: 218-147-6
FN: 1993
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Solution
Titration with HClO4 0.09 to 0.11 M
Melting point 52 to 56 °C
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR