2-Metylsyklohexanon 98%

Leverandør: Thermo Fisher Scientific
Warning

126610500 126612500 126610010
ACRO126610500EA 565 NOK
ACRO126610500 ACRO126612500 ACRO126610010
2-Metylsyklohexanon 98%
2-Metylsyklohexanon
Formel: C₇H₁₂O
Kokepunkt: 162…163 °C (760 mmHg)
Smeltepunkt: –14 °C
Tetthet: 0,924 g/cm³
Flash punkt: 48 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001635
CAS-nummer: 583-60-8
EINECS: 209-513-6
FN: 2297
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4500 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR