2-Nitro-p-toluidine 99%

Leverandør: Thermo Fisher Scientific
Danger

127240050 127241000 127245000
ACRO127240050EA 390 NOK
ACRO127240050 ACRO127241000 ACRO127245000
2-Nitro-p-toluidine 99%
2-Nitro-p-toluidine
Formel: C₇H₈N₂O₂
Kokepunkt: 169 °C (21 mmHg)
Smeltepunkt: 112…115 °C
Tetthet: 1,164 g/cm³ (20 °C)
Flash punkt: 157 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007907
CAS-nummer: 89-62-3
EINECS: 201-924-9
FN: 2660
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to orange-brown Crystalline powder
Infrared spectrum Conforms
Melting point 112 to 116 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR