3'-Nitroacetophenone 98%

Leverandør: Thermo Fisher Scientific

128331000 128335000
ACRO128331000EA 395 NOK
ACRO128331000 ACRO128335000
3'-Nitroacetophenone 98%
3'-Nitroacetophenone
Formel: C₈H₇NO₃
Kokepunkt: 202 °C (1013 hPa)
Smeltepunkt: 76…79 °C
Tetthet: 1,433 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007259
CAS-nummer: 121-89-1
EINECS: 204-504-3

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to yellow-green to yellow-brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR