Quinoxaline 98+%

Leverandør: Thermo Fisher Scientific
Warning

132221000
ACRO132221000EA 1450 NOK
ACRO132221000
Quinoxaline 98+%
Quinoxaline
Formel: C₈H₆N₂
Kokepunkt: 220…223 °C (760 mmHg)
Smeltepunkt: 29…34 °C
Tetthet: 1,1485 g/cm³ (32 °C)
Flash punkt: 98 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006719
CAS-nummer: 91-19-0
EINECS: 202-047-4
Merck Indeks: 12,08263

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to brown Low melting crystalline solid
Infrared spectrum Conforms
Melting point 29 to 34 °C
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR