D(-)-Ribose 99+%

Leverandør: Thermo Fisher Scientific

132360250 132361000 132360050 132365000
ACRO132360250EA 1150 NOK
ACRO132360250 ACRO132361000 ACRO132360050 ACRO132365000
D(-)-Ribose 99+%
D(-)-Ribose
Formel: C₅H₁₀O₅
Kokepunkt: 331 °C (1013 hPa)
Smeltepunkt: 88…92 °C
Tetthet: 0,8 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00135453
CAS-nummer: 50-69-1
EINECS: 200-059-4
Merck Indeks: 12,08371

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (40°C, 5 h, 20 Torr)
Sulfated ash ≤0.1 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -19.2° to -20.8° (20°C, 589 nm) (c=4, H2O)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR