1,3-Diphenyl-2-thiourea 98%

Leverandør: Thermo Fisher Scientific
Warning

138700010 138701000 138700025
ACRO138700010EA 1800 NOK
ACRO138700010 ACRO138701000 ACRO138700025
1,3-Diphenyl-2-thiourea 98%
1,3-Diphenyl-2-thiourea
Formel: C₁₃H₁₂N₂S
Kokepunkt: 365 °C (1013 hPa)
Smeltepunkt: 151…154 °C
Tetthet: 1,3205 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004921
CAS-nummer: 102-08-9
EINECS: 203-004-2
FN: 3077
ADR: 9,III
Merck Indeks: 13,03366

Bestill nå

Spesifikasjon Testresultater

Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 154 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR