Trifenylfosfin 99%

Leverandør: Thermo Fisher Scientific
Warning

140420010 140420250 140422500 140420050
ACRO140420010EA 2950 NOK
ACRO140420010 ACRO140420250 ACRO140422500 ACRO140420050
Trifenylfosfin 99%
Trifenylfosfin
Formel: C₁₈H₁₅P
Kokepunkt: 377 °C (760 mmHg)
Tetthet: 1,194 g/cm³ (20 °C)
Flash punkt: 181 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003043
CAS-nummer: 603-35-0
EINECS: 210-036-0
Merck Indeks: 12,09873

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 78.5 to 81.5 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR