D(+)-Xylose 99+%

Leverandør: Thermo Fisher Scientific

141001000 141005000 141000025
ACRO141001000EA 1350 NOK
ACRO141001000 ACRO141005000 ACRO141000025
D(+)-Xylose 99+%
D(+)-Xylose
Formel: C₅H₁₀O₅
Kokepunkt: 331 °C (1013 hPa)
Smeltepunkt: 148…153 °C
Tetthet: 1,525 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00151475
CAS-nummer: 58-86-6
EINECS: 200-400-7
Merck Indeks: 13,10142
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤0.2 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation +18.0° to +21.0° (20°C, 589 nm) (c=10, H2O/NH3)
Heavy metals (as Pb) ≤10 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR