EDTA dinatrium salt dihydrat 99+% for analysis

Leverandør: Thermo Fisher Scientific
Warning

147850010 147851000 147855000 147850050
ACRO147850010EA 2850 NOK
ACRO147850010 ACRO147851000 ACRO147855000 ACRO147850050
EDTA dinatrium salt dihydrat 99+% for analysis
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 252 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay 99.0 to 101.0 % (On dry substance)
Loss on drying ≤11.4 % (200°C, 3 h)
pH 4.0 to 6.0 (1 % at 20°C)
Iron (Fe) ≤80 ppm
Heavy metals (as Pb) ≤10 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR