Nicotinic acid hydrazide 97%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide

148200250 148201000
ACRO148200250EA 670 NOK
ACRO148200250 ACRO148201000
Nicotinic acid hydrazide 97%
Nicotinic acid hydrazide
Formel: C₆H₇N₃O
MW: 137.14 g/mol
Smeltepunkt: 160…163 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006383
CAS-nummer: 553-53-7
EINECS: 209-041-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Crystalline powder or needles
Infrared spectrum Conforms
Melting point 160 to 163 °C
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR