Dimethyl succinate 99%

Leverandør: Thermo Fisher Scientific
Warning

150731000 150735000
ACRO150731000EA 335 NOK
ACRO150731000 ACRO150735000
Dimethyl succinate 99%
Dimethyl succinate
Formel: C₆H₁₀O₄
Kokepunkt: 200 °C (1013 hPa)
Smeltepunkt: 18…19 °C
Tetthet: 1,11 g/cm³
Flash punkt: 85 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008466
CAS-nummer: 106-65-0
EINECS: 203-419-9
Merck Indeks: 12,09038

Bestill nå

Spesifikasjon Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4210 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤25 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR