Fluoroacetone 97%

Leverandør: Thermo Fisher Scientific
Danger

151780010 151780050
ACRO151780010EA 1225 NOK
ACRO151780010 ACRO151780050
Fluoroacetone 97%
Fluoroacetone
Formel: C₃H₅FO
Kokepunkt: 75 °C (760 mmHg)
Tetthet: 1,054 g/cm³
Flash punkt: 7 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00000451
CAS-nummer: 430-51-3
EINECS: 207-064-0
FN: 1992
ADR: 3,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3660 to 1.3700 (20°C, 589 nm)
GC ≥96.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR